Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026977.2 | NP_081253.1 | 3 | 428 | ||
GenBank mRNA | AK029090.1 | - | 3 | 428 | ||
AK145962.1 | - | 4 | 675 | |||
AK159252.1 | - | 150 | ||||
AK169779.1 | - | 3 | 700 | |||
BC013714.1 | - | 2 | 201 | |||
BC028815.1 | - | 3 | 413 | 5126467 |