Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAA2-SAA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG616641.1 | - | 6 | 532 |
Gene Symbol: SAA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK312182.1 | - | 4 | 336 | ||
BC007026.1 | - | 4 | 317 | 4071711 | ||
BG569210.1 | - | 4 | 386 | |||
CB113387.1 | - | 4 | 358 | |||
CR541758.1 | - | 3 | 260 | |||
M81349.1 | - | 4 | 335 |