Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026987.2 | NP_081263.2 | 19 | 3126 | ||
GenBank mRNA | AK010431.1 | - | 6 | 744 | ||
AK122314.2 | - | 20 | 3156 | |||
BC009147.1 | - | 19 | 3085 | 2647184 | ||
BC046781.1 | - | 19 | 3054 | 5716182 | ||
BC138204.1 | - | 19 | 3065 | 8861223 | ||
BC138205.1 | - | 19 | 3065 | 8861224 |