Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093752.1 | NP_001087221.1 | 5 | 646 | ||
NM_001093753.1 | NP_001087222.1 | 4 | 628 | |||
GenBank mRNA | AK011472.1 | - | 5 | 584 | ||
AK029394.1 | - | 4 | 591 | |||
AK081511.1 | - | 2 | 453 | |||
AK146719.1 | - | 4 | 600 | |||
AK183987.1 | - | 3 | 328 | |||
AK216204.1 | - | 3 | 329 | |||
AK218039.1 | - | 3 | 329 | |||
AK218604.1 | - | 3 | 218 | |||
BC002111.1 | - | 4 | 599 | |||
BC021809.1 | - | 4 | 583 | |||
BC037053.1 | - | 5 | 569 | |||
BC038928.1 | - | 5 | 605 | |||
BC055763.1 | - | 6 | 534 | |||
BC069945.1 | - | 4 | 578 | |||
BC138145.1 | - | 2 | 381 | 8861164 | ||
BC145162.1 | - | 2 | 381 | 9053713 |