Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027052.3 | NP_081328.2 | 3 | 315 | ||
GenBank mRNA | AB055004.1 | - | 3 | 291 | ||
AK051023.1 | - | 3 | 316 | |||
AK164292.1 | - | 3 | 258 | |||
AK167539.1 | - | 3 | 297 | |||
AK169349.1 | - | 2 | 304 | |||
AY027919.1 | - | 3 | 354 | |||
BC024072.1 | - | 3 | 240 | 5097293 | ||
BC024123.1 | - | 3 | 272 | 5098067 | ||
BC031717.1 | - | 3 | 233 | 4195569 |