Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027081.3 | NP_081357.2 | 1, 2 | 264 | ||
GenBank mRNA | AK084555.1 | - | 1, 2 | 257 | ||
AK154949.1 | - | 1, 2 | 286 | |||
BB658928.1 | - | 1, 2 | 257 | |||
BC139003.1 | - | 1, 2 | 238 | 8862025 | ||
BY252285.1 | - | 1, 2 | 266 | |||
BY748566.1 | - | 1, 2 | 281 |