Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027081.3 | NP_081357.2 | 20 | 1995 | ||
GenBank mRNA | AK084555.1 | - | 18 | 2022 | ||
AK154949.1 | - | 16 | 2648 | |||
BC017544.1 | - | 12 | 1951 | |||
BC030395.1 | - | 18 | 2667 | |||
BC058958.1 | - | 16 | 1502 | |||
BC080797.1 | - | 16 | 1884 | |||
BC139003.1 | - | 20 | 1969 | 8862025 |