Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122873.1 | NP_001116345.1 | 7 | 755 | ||
NM_027086.3 | NP_081362.2 | 7 | 703 | |||
GenBank mRNA | AK009025.1 | - | 7 | 704 | ||
AK154496.1 | - | 7 | 713 | |||
AK179100.1 | - | 3 | 267 | |||
AK182178.1 | - | 3 | 266 | |||
AK183337.1 | - | 3 | 267 | |||
AK207226.1 | - | 3 | 267 | |||
AK209878.1 | - | 3 | 287 | |||
AK212304.1 | - | 3 | 268 | |||
AK214469.1 | - | 1 | 115 | |||
BC016456.1 | - | 6 | 689 | 4912715 | ||
BC094047.1 | - | 6 | 692 | 5253101 | ||
CX203425.1 | - | 7 | 730 |