Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027093.4 | NP_081369.2 | 11 | 1048 | ||
GenBank mRNA | AK009123.1 | - | 11 | 1115 | ||
BC005630.1 | - | 11 | 915 |