Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027102.3 | NP_081378.1 | 2, 3 | 390 | ||
GenBank mRNA | AF361882.1 | - | 2, 3 | 375 | ||
AK009223.1 | - | 2, 3 | 366 | |||
AK156399.1 | - | 2, 3 | 388 | |||
BC019460.1 | - | 2, 3 | 374 | 4188050 | ||
BC049249.1 | - | 2, 3 | 370 | 5039892 | ||
BQ043033.1 | - | 2, 3 | 331 | |||
CJ048383.1 | - | 2, 3 | 372 |