Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027113.2 | NP_081389.1 | 1 | 533 | ||
GenBank mRNA | AK009247.1 | - | 1 | 448 | ||
AK149554.1 | - | 504 | ||||
AK165100.1 | - | 397 | ||||
BC064045.1 | - | 1 | 533 | 1245342 | ||
EF011616.1 | - | 1 | 531 |