Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291031.1 | NP_001277960.1 | 3 | 1511 | ||
NM_001291032.1 | NP_001277961.1 | 4 | 1078 | |||
NM_027129.3 | NP_081405.2 | 4 | 1422 | |||
GenBank mRNA | AK002807.1 | - | 3 | 1431 | ||
AK009634.1 | - | 4 | 1015 | |||
BC048399.1 | - | 4 | 1342 | 6438951 |