Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027193.2 | NP_081469.2 | 5, 6 | 602 | ||
GenBank mRNA | AK010475.1 | - | 5, 6 | 592 | ||
AK034652.1 | - | 612 | ||||
BC060372.1 | - | 5, 6 | 602 | 6391546 |