Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027194.3 | NP_081470.2 | 2 | 397 | ||
GenBank mRNA | AK010545.1 | - | 2 | 329 | ||
AK045768.1 | - | 2 | 356 | |||
AK080854.1 | - | 2 | 325 | |||
AK083654.1 | - | 2 | 318 | |||
AK085975.1 | - | 2 | 349 | |||
AK086015.1 | - | 2 | 335 | |||
AK086059.1 | - | 2 | 351 | |||
AK088246.1 | - | 2 | 349 | |||
AK149407.1 | - | 2 | 326 | |||
AK160039.1 | - | 2 | 334 | |||
AK166012.1 | - | 2 | 350 | |||
BC026789.1 | - | 2 | 324 | 4188168 |