Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006544.3 | NP_006535.1 | 3 | 412 | ||
XM_005267272.3 | XP_005267329.1 | 3 | 497 | |||
GenBank mRNA | AK303531.1 | - | 3 | 350 | ||
AK312523.1 | - | 3 | 417 | |||
BC041126.1 | - | 3 | 378 | 3914527 | ||
BI460915.1 | - | 3 | 423 | |||
BI461191.1 | - | 3 | 423 | |||
U85946.1 | - | 3 | 356 |