Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027211.2 | NP_081487.1 | 5, 6 | 434 | ||
GenBank mRNA | AJ306451.1 | - | 5, 6 | 434 | ||
BB617542.1 | - | 5, 6 | 431 | |||
BC013521.1 | - | 5, 6 | 404 | 4236501 |