Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253694.1 | NP_001240623.1 | 4 | 935 | ||
NM_001253695.1 | NP_001240624.1 | 3 | 664 | |||
NM_001253696.1 | NP_001240625.1 | 2 | 733 | |||
NM_027248.2 | NP_081524.2 | 3 | 822 | |||
GenBank mRNA | AB063578.1 | - | 3 | 822 | ||
AK008516.1 | - | 1 | 140 | |||
AK053445.1 | - | 3 | 659 | |||
AK154332.1 | - | 4 | 910 | |||
BC034874.1 | - | 3 | 698 | |||
BC052050.1 | - | 4 | 708 | |||
BC071271.1 | - | 2 | 593 | 30290129 | ||
BC096486.1 | - | 2 | 701 | 5352045 | ||
BQ890732.1 | - | 1 | 458 |