Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253694.1 | NP_001240623.1 | 6 | 2600 | ||
NM_001253695.1 | NP_001240624.1 | 5 | 2329 | |||
NM_001253696.1 | NP_001240625.1 | 4 | 2398 | |||
NM_027248.2 | NP_081524.2 | 5 | 2487 | |||
GenBank mRNA | AB063578.1 | - | 5 | 2487 | ||
AK053445.1 | - | 5 | 2326 | |||
AK154332.1 | - | 6 | 2575 | |||
BC010447.1 | - | 3 | 937 | |||
BC034874.1 | - | 4 | 1468 | |||
BC052050.1 | - | 6 | 2373 | |||
BC071271.1 | - | 4 | 2258 | 30290129 | ||
BC096486.1 | - | 4 | 2366 | 5352045 |