Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027260.3 | NP_081536.2 | 11, 12 | 1356 | ||
GenBank mRNA | AF513620.1 | - | 10, 11 | 1020 | ||
AK012664.1 | - | 11, 12 | 1048 | |||
AK154485.1 | - | 11, 12 | 1069 |