Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027276.2 | NP_081552.2 | 17, 18 | 1719 | ||
GenBank mRNA | AK013213.1 | - | 6, 7 | 653 | ||
AK147938.1 | - | 17, 18 | 1734 | |||
AK167965.1 | - | 17, 18 | 1734 | |||
AK168978.1 | - | 17, 18 | 1706 | |||
AK181696.1 | - | 3, 4 | 301 | |||
AK184248.1 | - | 3, 4 | 291 | |||
AK188605.1 | - | 3, 4 | 295 | |||
AK198756.1 | - | 2, 3 | 152 | |||
AK205790.1 | - | 3, 4 | 277 | |||
AK214776.1 | - | 3, 4 | 248 | |||
BC005658.1 | - | 1, 2 | 316 | |||
BC026606.1 | - | 17, 18 | 1719 | 4976424 |