Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142950.1 | NP_001136422.1 | 4, 5 | 392 | ||
NM_027350.3 | NP_081626.2 | 4, 5 | 389 | |||
GenBank mRNA | AK077699.1 | - | 4, 5 | 370 | ||
AK083730.1 | - | 4, 5 | 349 | |||
AK157244.1 | - | 4, 5 | 351 | |||
AK172657.1 | - | 4, 5 | 351 | |||
AK191424.1 | - | 3, 4 | 210 | |||
BC005473.1 | - | 4, 5 | 295 | |||
BC052849.1 | - | 4, 5 | 350 | 30057822 |