Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142950.1 | NP_001136422.1 | 11 | 1255 | ||
NM_027350.3 | NP_081626.2 | 11 | 1252 | |||
GenBank mRNA | AK013880.1 | - | 2 | 232 | ||
AK077699.1 | - | 11 | 1233 | |||
AK157244.1 | - | 11 | 1215 | |||
AK172657.1 | - | 11 | 1214 | |||
AK195743.1 | - | 1 | 113 | |||
BC005473.1 | - | 11 | 1158 | |||
BC052849.1 | - | 11 | 1213 | 30057822 |