Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027352.4 | NP_081628.3 | 10 | 1578 | ||
GenBank mRNA | AK002515.1 | - | 11 | 1656 | ||
AK033413.1 | - | 10 | 1512 | |||
AK077153.1 | - | 10 | 1453 | |||
AK077683.1 | - | 10 | 1513 | |||
AK077706.1 | - | 10 | 1445 | |||
AK088239.1 | - | 10 | 1459 | |||
AK148337.1 | - | 10 | 1512 | |||
AK151843.1 | - | 10 | 1460 | |||
AK152475.1 | - | 10 | 1460 | |||
AK152544.1 | - | 10 | 1460 | |||
AK152603.1 | - | 10 | 1488 | |||
AK153049.1 | - | 10 | 1460 | |||
AK153555.1 | - | 10 | 1511 | |||
AK159469.1 | - | 10 | 1510 | |||
AK167823.1 | - | 10 | 1512 | |||
AK168928.1 | - | 11 | 1708 | |||
AK169382.1 | - | 10 | 1460 | |||
BC005600.1 | - | 10 | 1451 | 3482776 | ||
BC007468.1 | - | 3 | 1159 | |||
BC016455.1 | - | 9 | 1303 | 4234343 | ||
BC024982.1 | - | 10 | 1473 |