Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285905.1 | NP_001272834.1 | 7 | 2565 | ||
NM_001285906.1 | NP_001272835.1 | 7 | 2579 | |||
NM_001285907.1 | NP_001272836.1 | 7 | 2707 | |||
NM_027353.4 | NP_081629.1 | 7 | 2559 | |||
GenBank mRNA | AK005198.1 | - | 1 | 84 | ||
AK030393.1 | - | 7 | 2441 | |||
AK034689.1 | - | 7 | 2483 | |||
AK154346.1 | - | 7 | 2444 | |||
AK173122.1 | - | 6 | 2401 | |||
BC006920.1 | - | 7 | 2475 | 3601764 | ||
BC079575.1 | - | 6 | 2634 |