Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027355.2 | NP_081631.2 | 1 | 800 | ||
GenBank mRNA | AK013946.1 | - | 1 | 760 | ||
AK053769.1 | - | 1 | 803 | |||
BC046815.1 | - | 1 | 772 | 5702430 |