Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190885.1 | NP_001177814.1 | 9 | 1346 | ||
NM_001190886.1 | NP_001177815.1 | 8 | 1302 | |||
NM_001290690.1 | NP_001277619.1 | 8 | 1501 | |||
NM_027398.3 | NP_081674.2 | 8 | 1313 | |||
GenBank mRNA | AK014313.1 | - | 8 | 1301 | ||
AK045948.1 | - | 9 | 1347 | |||
AK046398.1 | - | 9 | 1335 | |||
AK081845.1 | - | 8 | 1303 | |||
AY171234.1 | - | 8 | 1377 | |||
BC034241.1 | - | 8 | 1255 | 4503251 | ||
DQ148493.1 | - | 8 | 1255 | |||
DQ148494.1 | - | 8 | 1303 |