Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027399.3 | NP_081675.2 | 3 | 214 | ||
GenBank mRNA | AF297098.1 | - | 2 | 144 | ||
AK010437.1 | - | 3 | 209 | |||
AK144312.1 | - | 3 | 169 | |||
AY029584.1 | - | 3 | 173 | |||
BC061023.1 | - | 3 | 199 | 30304473 | ||
DN176485.1 | - | 3 | 214 |