Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027411.2 | NP_081687.2 | 2 | 294 | ||
GenBank mRNA | AK011138.1 | - | 2 | 266 | ||
AK012927.1 | - | 2 | 295 | |||
AK139553.1 | - | 2 | 170 | |||
AK163303.1 | - | 2 | 246 | |||
BC006674.1 | - | 2 | 277 | 3499487 | ||
BY712013.1 | - | 2 | 295 |