Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810408A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027419.3 | NP_081695.2 | 6 | 1297 | ||
GenBank mRNA | AK013042.1 | - | 6 | 1298 | ||
AK015537.1 | - | 6 | 1223 | |||
AK051029.1 | - | 6 | 1385 | |||
AK160144.1 | - | 6 | 1192 | |||
AK171674.1 | - | 6 | 1344 | |||
BC005538.1 | - | 5 | 589 | |||
BC051546.1 | - | 6 | 1259 | 5255109 | ||
BC055710.1 | - | 6 | 1402 | |||
BC069874.1 | - | 6 | 1297 | 4216824 |