Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027425.3 | NP_081701.2 | 7 | 747 | ||
GenBank mRNA | AK011847.1 | - | 7 | 717 | ||
AK083205.1 | - | 7 | 732 | |||
AK173195.1 | - | 7 | 708 | |||
BC017645.1 | - | 7 | 709 | |||
BC137736.1 | - | 7 | 696 | 8860755 | ||
BC137737.1 | - | 7 | 696 | 8860756 |