Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ810302.1 | - | 8 | 1340 | ||
AJ810303.1 | - | 8 | 1212 | |||
AK293696.1 | - | 7 | 1197 | |||
BC017807.1 | - | 8 | 1353 | 4690513 |