Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027727.2 | NP_082003.1 | 9 | 1191 | ||
GenBank mRNA | AK016957.1 | - | 9 | 1119 | ||
AK049909.1 | - | 9 | 1156 | |||
AK140643.1 | - | 1 | 296 | |||
AK149628.1 | - | 9 | 1165 | |||
AK149891.1 | - | 9 | 1165 | |||
AK207614.1 | - | 1 | 136 | |||
AK215313.1 | - | 2 | 279 | |||
BC018220.1 | - | 5 | 551 | 3985017 | ||
BC038276.1 | - | 9 | 1193 | 4458792 |