Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029573.2 | NP_083849.1 | 10 | 987 | ||
GenBank mRNA | AK003393.1 | - | 7 | 880 | ||
AK010065.1 | - | 10 | 1011 | |||
AK032787.1 | - | 10 | 1011 | |||
AK047951.1 | - | 10 | 1009 | |||
AK117103.1 | - | 10 | 1023 | |||
AK146717.1 | - | 7 | 851 | |||
AK150618.1 | - | 10 | 1018 | |||
AK151304.1 | - | 10 | 1012 | |||
AK152353.1 | - | 10 | 1021 | |||
AK153459.1 | - | 10 | 1012 | |||
AK158646.1 | - | 10 | 1165 | |||
AK159051.1 | - | 10 | 1084 | |||
AK159666.1 | - | 10 | 1016 | |||
AK160054.1 | - | 10 | 1016 | |||
AK164038.1 | - | 11 | 1114 | |||
AK167822.1 | - | 10 | 1017 | |||
AK168034.1 | - | 10 | 1007 | |||
AK168049.1 | - | 10 | 1012 | |||
AK168149.1 | - | 10 | 1018 | |||
AK169152.1 | - | 10 | 985 | |||
AK196914.1 | - | 1 | 101 | |||
BC034273.1 | - | 10 | 977 | 4166193 | ||
BC049956.1 | - | 10 | 987 | 4212315 |