Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030566.2 | NP_085043.2 | 8 | 1449 | ||
GenBank mRNA | AK029188.1 | - | 6 | 807 | ||
AK089938.1 | - | 8 | 1153 | |||
AK144836.1 | - | 8 | 1135 | |||
AK157832.1 | - | 8 | 1110 | |||
BC004088.1 | - | 7 | 1195 | 3593722 | ||
BC015287.1 | - | 8 | 1457 | 4240442 |