Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103181.1 | NP_001096651.1 | 11 | 1717 | ||
NM_054040.3 | NP_473381.1 | 11 | 2559 | |||
GenBank mRNA | AF219945.1 | - | 11 | 1963 | ||
AK053530.1 | - | 4 | 380 | |||
AK147643.1 | - | 11 | 1657 | |||
AK173165.1 | - | 11 | 2559 | |||
AK207024.1 | - | 3 | 294 | |||
BC138024.1 | - | 11 | 1894 | 8861043 | ||
BC138025.1 | - | 11 | 1894 | 8861044 |
Gene Symbol: Gm29719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049966.1 | - | 1661 |