Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080555.2 | NP_542122.1 | 6 | 1514 | ||
GenBank mRNA | AK011276.1 | - | 6 | 1512 | ||
AK076183.1 | - | 6 | 1210 | |||
AK159136.1 | - | 6 | 1600 | |||
AK160056.1 | - | 6 | 1414 | |||
AK163427.1 | - | 1 | 14 | |||
BC005558.1 | - | 6 | 1501 | 3964817 |