Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177998.1 | NP_001171469.1 | 4, 5 | 465 | ||
NM_001177999.1 | NP_001171470.1 | 4, 5 | 488 | |||
NM_006424.2 | NP_006415.2 | 4, 5 | 468 | |||
GenBank mRNA | AF111856.1 | - | 4, 5 | 450 | ||
AF146796.1 | - | 4, 5 | 419 | |||
AK075046.1 | - | 4, 5 | 430 | |||
AK292675.1 | - | 4, 5 | 456 | |||
AK314514.1 | - | 4, 5 | 477 | |||
BC142704.1 | - | 4, 5 | 413 | 40148805 | ||
BC146666.1 | - | 4, 5 | 413 | 40148124 | ||
DB201517.1 | - | 4, 5 | 465 | |||
DB241180.1 | - | 4, 5 | 488 | |||
KM822236.1 | - | 3, 4 | 387 |