Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080556.3 | NP_542123.3 | 1, 2 | 353 | ||
GenBank mRNA | AK010325.1 | - | 1, 2 | 310 | ||
AK030340.1 | - | 1, 2 | 283 | |||
AK048012.1 | - | 1, 2 | 272 | |||
AK050338.1 | - | 1, 2 | 300 | |||
AK075720.1 | - | 1, 2 | 353 | |||
AK088905.1 | - | 1, 2 | 325 | |||
AK159757.1 | - | 1, 2 | 298 | |||
AK209133.1 | - | 40 | ||||
AK219057.1 | - | 40 | ||||
BB619330.1 | - | 1, 2 | 354 | |||
BC003862.1 | - | 1, 2 | 292 | 3488326 |