Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164600.1 | NP_001158072.1 | 11 | 1497 | ||
NM_028364.2 | NP_082640.2 | 12 | 1578 | |||
NM_133691.5 | NP_598452.2 | 11 | 1527 | |||
GenBank mRNA | AK012434.1 | - | 11 | 1465 | ||
AK075960.1 | - | 11 | 1495 | |||
AK146534.1 | - | 12 | 1549 | |||
AK149632.1 | - | 12 | 1544 | |||
AK151526.1 | - | 11 | 1491 | |||
AK168789.1 | - | 11 | 1494 | |||
BC010601.1 | - | 11 | 1497 | 3257153 |