Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133693.2 | NP_598454.2 | 2, 3 | 419 | ||
GenBank mRNA | AK013929.1 | - | 2, 3 | 357 | ||
AK151000.1 | - | 2, 3 | 331 | |||
AK185695.1 | - | 1, 2 | 25 | |||
AK194325.1 | - | 1, 2 | 25 | |||
AK197356.1 | - | 1, 2 | 25 | |||
AK210537.1 | - | 1, 2 | 25 | |||
AK211257.1 | - | 1, 2 | 72 | |||
AK213653.1 | - | 1, 2 | 72 | |||
AK216265.1 | - | 1, 2 | 72 | |||
BC009148.1 | - | 2, 3 | 356 | 3600406 | ||
BC011286.1 | - | 2, 3 | 338 | 3485410 | ||
BC027372.1 | - | 2, 3 | 343 | 4952304 | ||
BC039917.1 | - | 2, 3 | 354 | |||
BC057928.1 | - | 2, 3 | 345 | 5289543 |