Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133698.2 | NP_598459.2 | 3 | 321 | ||
GenBank mRNA | AK041026.1 | - | 3 | 322 | ||
AY027660.1 | - | 284 | ||||
BC118008.1 | - | 2 | 284 | 40087781 |