Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133698.2 | NP_598459.2 | 3 | 591 | ||
GenBank mRNA | AY027660.1 | - | 554 | |||
BC118008.1 | - | 2 | 554 | 40087781 |