Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdspl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133710.3 | NP_598471.3 | 3, 4 | 600 | ||
GenBank mRNA | AA239542.1 | - | 3, 4 | 291 | ||
AJ344340.1 | - | 3, 4 | 682 | |||
AK013115.1 | - | 3, 4 | 596 | |||
BC094289.1 | - | 3, 4 | 285 | 4986338 |