Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133718.4 | NP_598479.1 | 5 | 813 | ||
GenBank mRNA | AK008408.1 | - | 5 | 699 | ||
AK077065.1 | - | 5 | 816 | |||
AK150071.1 | - | 5 | 716 | |||
AK159364.1 | - | 5 | 740 | |||
AK161096.1 | - | 5 | 695 | |||
AK170171.1 | - | 5 | 716 | |||
AK170697.1 | - | 5 | 717 | |||
BC018367.1 | - | 5 | 665 | 2648510 | ||
BC026136.1 | - | 4 | 289 |