Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133718.4 | NP_598479.1 | 7 | 3542 | ||
GenBank mRNA | AK077065.1 | - | 7 | 3545 | ||
AK078136.1 | - | 1 | 854 | |||
AK150071.1 | - | 7 | 3445 | |||
AK170697.1 | - | 7 | 3447 | |||
BC018367.1 | - | 7 | 3395 | 2648510 | ||
BC018491.1 | - | 7 | 3422 | 2651602 | ||
BC026136.1 | - | 6 | 3017 | |||
BY470970.1 | - | 1 | 358 |