Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cysltr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162412.1 | NP_001155884.1 | 5 | 1724 | ||
NM_133720.3 | NP_598481.2 | 6 | 1781 | |||
GenBank mRNA | AF331658.1 | - | 6 | 1740 | ||
AK008997.1 | - | 5 | 1719 | |||
BE848050.1 | - | 1 | 236 |
Set Membership: |
G-protein coupled receptor |