Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133729.1 | NP_598490.1 | 7 | 939 | ||
GenBank mRNA | AK011445.1 | - | 7 | 1323 | ||
AK029278.1 | - | 5 | 1544 | |||
AK034609.1 | - | 7 | 978 | |||
BC005708.1 | - | 7 | 939 | 2598933 | ||
BC132438.1 | - | 6 | 742 | 40130715 | ||
BC138193.1 | - | 6 | 742 | 8861212 | ||
BC145189.1 | - | 6 | 742 |