Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133950.2 | NP_598711.1 | 5 | 1242 | ||
GenBank mRNA | AJ278132.1 | - | 5 | 1242 | ||
AK013118.1 | - | 5 | 1066 | |||
AK078785.1 | - | 5 | 1311 | |||
AK188469.1 | - | 1 | 48 | |||
BC011370.1 | - | 5 | 1154 | 3152867 |