Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134255.3 | NP_599016.2 | 8 | 1063 | ||
GenBank mRNA | AK028761.1 | - | 8 | 1003 | ||
AK031743.1 | - | 8 | 1066 | |||
AK045274.1 | - | 8 | 1042 | |||
AK051580.1 | - | 8 | 1060 | |||
AK085663.1 | - | 8 | 1026 | |||
AK085696.1 | - | 8 | 1013 | |||
AK088153.1 | - | 8 | 1001 | |||
AK162451.1 | - | 8 | 1064 | |||
AK167755.1 | - | 8 | 1004 | |||
AK168360.1 | - | 8 | 1004 | |||
AK168987.1 | - | 8 | 990 | |||
BC022911.1 | - | 8 | 972 | 4457285 |