Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134255.3 | NP_599016.2 | 8 | 1680 | ||
GenBank mRNA | AK004319.1 | - | 1 | 275 | ||
AK028761.1 | - | 8 | 1619 | |||
AK031743.1 | - | 8 | 1683 | |||
AK045274.1 | - | 8 | 1659 | |||
AK051580.1 | - | 8 | 1678 | |||
AK085663.1 | - | 8 | 1643 | |||
AK085696.1 | - | 8 | 1630 | |||
AK088153.1 | - | 8 | 1619 | |||
AK162451.1 | - | 8 | 1681 | |||
AK167755.1 | - | 8 | 1621 | |||
AK168360.1 | - | 8 | 1621 | |||
AK168987.1 | - | 8 | 1608 | |||
AK207677.1 | - | 1 | 193 | |||
BC022911.1 | - | 8 | 1589 | 4457285 |